Learn More
Labcon North America Eclipse™ 300uL Pipet Tips, Individually Wrapped , Sterile
Catalog No. 50101678
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 300uL Pipettors ; Beveled Point; Sterile; Individually Wrapped; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Yellow color; 1 tip per bag; 200 tips per pack; 10 packs per case; 52.1mm. H; Made in USA
Catalog No. 50-101-678 Supplier Labcon North America Supplier No. 10391650009
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 300uL Pipettors ; Beveled Point; Sterile; Individually Wrapped; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Yellow color; 1 tip per bag; 200 tips per pack; 10 packs per case; 52.1mm. H; Made in USA