Learn More
Labcon North America Eclipse™ 200uL Clear Pipet Tips, in Resealable Bags
Catalog No. 50101685
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Clear; 1000 tips per bag; 10 bags per case; 49.7mm. H; Made in USA
Catalog No. 50-101-685 Supplier Labcon North America Supplier No. 10608000009
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Clear; 1000 tips per bag; 10 bags per case; 49.7mm. H; Made in USA