Learn More
Labcon North America Eclipse™ 200uL Graduated Pipet Tips, in 96 Racks, Sterile
Catalog No. 50101643
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Graduated; Fits Most Popular 200uL Pipettors ; Beveled Point; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Clear; 96 tips per rack; 10 racks per pack; 10 packs per case; 49.7mm. H; Made in USA
Catalog No. 50-101-643 Supplier Labcon North America Supplier No. 10939650089
Tip, Pipet; Labcon; Eclipse™; Graduated; Fits Most Popular 200uL Pipettors ; Beveled Point; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Clear; 96 tips per rack; 10 racks per pack; 10 packs per case; 49.7mm. H; Made in USA