Learn More
Labcon North America Eclipse™ FlexTop™ 1250uL Extended Length Pipet Tips with UltraFine™ points, in Resealable Bags
Catalog No. 50101639
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extended Length; Fits popular 1000uL and 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 88.9mm. H; Made in USA
Catalog No. 50-101-639 Supplier Labcon North America Supplier No. 10458000009
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extended Length; Fits popular 1000uL and 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 1000 tips/bag; 10 bags per case; 88.9mm. H; Made in USA