Learn More
Labcon North America Eclipse™ FlexTop™ 1250uL Extended Length Pipet Tips with UltraFine™ points, in 96 Racks
Catalog No. 50101641
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extended Length; Fits popular 1000 and 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks/pack; 8 packs per case; 88.9mm. H; Made in USA
Catalog No. 50-101-641 Supplier Labcon North America Supplier No. 10459600089
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extended Length; Fits popular 1000 and 1250uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks/pack; 8 packs per case; 88.9mm. H; Made in USA