Learn More
Labcon North America Eclipse™ FlexTop™ 1250uL Extended Length Pipet Tips with UltraFine™ points, Individually Wrapped, Sterile
Catalog No. 50101640
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extended Length; Fits Most Popular 1000 and 1250uL Pipettors ; Sterile; Individually Wrapped; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Clear; 1 tip per bag; 200 tips per pack; 10 packs per case; 88.9mm. H; Made in USA
Catalog No. 50-101-640 Supplier Labcon North America Supplier No. 10451650009
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extended Length; Fits Most Popular 1000 and 1250uL Pipettors ; Sterile; Individually Wrapped; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Clear; 1 tip per bag; 200 tips per pack; 10 packs per case; 88.9mm. H; Made in USA