Learn More
Labcon North America Eclipse™ 200uL Yellow Pipet Tips, in Pagoda Express Refills
Catalog No. 50101698
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Yellow color; 960 tips per Pagoda Express refill; 10 refills per case; 49.7mm. H; Made in USA
Catalog No. 50-101-698 Supplier Labcon North America Supplier No. 10302900209
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Yellow color; 960 tips per Pagoda Express refill; 10 refills per case; 49.7mm. H; Made in USA