Learn More
Labcon North America Eclipse™ 200uL Yellow Pipet Tips, in 192 Stack Rack Refills
Catalog No. 50101690
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Yellow color; 192 tips per rack; 5 racks per stack ; 10 stacks per case; 49.7mm. H; Made in USA
Catalog No. 50-101-690 Supplier Labcon North America Supplier No. 10307000289
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Yellow color; 192 tips per rack; 5 racks per stack ; 10 stacks per case; 49.7mm. H; Made in USA