Learn More
Labcon North America Eclipse™ 200uL Yellow Pipet Tips, in Pagoda Refills, Sterile
Catalog No. 50101697
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Yellow color; 960 tips per Pagodarefill (includes 1 free refillable rack); 10 refills per case; 49.7mm. H; Made in USA
Catalog No. 50-101-697 Supplier Labcon North America Supplier No. 10302950089
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; Sterile; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Yellow color; 960 tips per Pagodarefill (includes 1 free refillable rack); 10 refills per case; 49.7mm. H; Made in USA