Learn More
Labcon North America Eclipse™ 200uL Yellow Pipet Tips, in 96 Racks
Catalog No. 50101694
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Yellow color; 96 tips per rack; 10 racks per pack; 10 packs per case; 49.7mm. H; Made in USA
Catalog No. 50-101-694 Supplier Labcon North America Supplier No. 10309600089
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; ; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Yellow color; 96 tips per rack; 10 racks per pack; 10 packs per case; 49.7mm. H; Made in USA