Learn More
Labcon North America 200uL Yellow Pipet Tips, Individually Wrapped, Sterile, 2000/cs (10 packs x 200 per pack)
Catalog No. 50101700
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; Sterile; Individually Wrapped; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Yellow color; 1 tip per bag; 200 tips per pack; 10 packs per case; 49.7mm. H; Made in USA
Catalog No. 50-101-700 Supplier Labcon North America Supplier No. 10301650009
Tip, Pipet; Labcon; Eclipse™; Fits Most Popular 200uL Pipettors ; Beveled Point; Sterile; Individually Wrapped; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Yellow color; 1 tip per bag; 200 tips per pack; 10 packs per case; 49.7mm. H; Made in USA