Learn More
Labcon North America Eclipse™ FlexTop™ 200uL Extended Pipet Tips with UltraFine™ points, in 96 Racks
Catalog No. 50101701
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extra long; Fits popular 200uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks per pack; 8 packs per case; 83.8mm. H; Made in USA
Catalog No. 50-101-701 Supplier Labcon North America Supplier No. 10129600089
Tip, Pipet; Labcon; Eclipse™ FlexTop™; Extra long; Fits popular 200uL pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 96 tips/rack; 6 racks per pack; 8 packs per case; 83.8mm. H; Made in USA