Learn More
Aviva Systems Biology Corp TIP120A Antibody - N-terminal region (ARP34585_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34585_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TIP120A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TIP120A. Alternate names: TIP120, TIP120AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55463