Learn More
Cayman Chemical TIP39 (human, bovine) (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502918189
Encompass_Preferred
A neuropeptide and PTH2R agonist; induces cAMP accumulation in COS-7 cells expressing recombinant human or rat PTH2R (EC50s = 0.5 and 0.8 nM, respectively) and F-11 cells that endogenously express PTH2R (EC50 = 1.15 nM); induces cell cycle arrest at the G0/G1 phase and decreases expression of Sox9 in CFK2 rat chondrocytes at 1 nM; decreases immobility time in the forced swim test in mice
Catalog No. 50-291-8189 Supplier Cayman Chemical Supplier No. 3745210 MG
A neuropeptide and PTH2R agonist; induces cAMP accumulation in COS-7 cells expressing recombinant human or rat PTH2R (EC50s = 0.5 and 0.8 nM, respectively) and F-11 cells that endogenously express PTH2R (EC50 = 1.15 nM); induces cell cycle arrest at the G0/G1 phase and decreases expression of Sox9 in CFK2 rat chondrocytes at 1 nM; decreases immobility time in the forced swim test in mice