Learn More
Aviva Systems Biology Corp TIPARP Antibody - N-terminal region (ARP33763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIPARP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TIPARP. Alternate names: DDF1, DKFZP434J214, DKFZp686N0351, DKFZp686P1838, FLJ40466, PARP-1, PARP-7, PARP7, pART14ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55141