Learn More
Biomatik Corporation Tiparp Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11508
Encompass_Preferred
Tiparp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TIPARP. The Tiparp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tcdd-Inducible Poly [Adp-Ribose] Polymerase Protein (74-178Aa). Applications: ELISA, WB, IHC. Target Synonyms: TIPARP. Accession Number: Q7Z3E1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6188