Learn More
ABclonal Technology TIPARP Rabbit pAb
SDP
Supplier:  ABclonal Technology A143291000UL
Encompass_Preferred
This gene encodes a member of the poly(ADP-ribose) polymerase superfamily. Studies of the mouse ortholog have shown that the encoded protein catalyzes histone poly(ADP-ribosyl)ation and may be involved in T-cell function. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-09207