Learn More
Medchemexpress LLC HY-121685A 5mg Medchemexpress, Tipepidine (hydrochloride) CAS:1449686-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY121685A5MG
Encompass_Preferred
Medchemexpress, HY-121685A 5mg Tipepidine (HCl) CAS:1449686-84-3 Tipepidine hydrochloride reversibly inhibits dopamine (DA) D2 receptor-mediated GIRK currents (IDA(GIRK)) with an IC50 of 7.0 μM. Tipepidine hydrochloride subsequently activates VTA dopamine neuron. Tipepidine hydrochloride, a non-narcotic antitussive, exerts an antidepressant-like effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9624