Learn More
Medchemexpress LLC HY-10502 10mg Medchemexpress, Tipifarnib CAS:192185-72-1 Purity:>98%
SDP
Catalog No. 502028763
Encompass_Preferred
Medchemexpress, HY-10502 10mg Tipifarnib CAS:192185-72-1 Tipifarnib (IND 58359) binds to and inhibits farnesyltransferase (FTase) with an IC50 of 0.86 nM. Antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8763 Supplier Medchemexpress LLC Supplier No. HY1050210MG
Medchemexpress, HY-10502 10mg Tipifarnib CAS:192185-72-1 Tipifarnib (IND 58359) binds to and inhibits farnesyltransferase (FTase) with an IC50 of 0.86 nM. Antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.