Learn More
Selleck Chemical LLC Tipifarnib 10mg 192185-72-1 R115777
SDP
Supplier:  Selleck Chemical LLC S145310MG
Encompass_Preferred
Tipifarnib (R115777) is a potent and specific farnesyltransferase (FTase) inhibitor with IC50 of 0.6 nM, its anti-proliferative effects are most prominent in H-ras or N-ras mutant cells. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-847-8