Learn More
Apexbio Technology LLC Tipifarnib (Zarnestra), 5mg. Cas: 192185-72-1 MFCD: MFCD07772347. Farnesyltransferase inhibitor,potent and specific
SDP
Supplier:  Apexbio Technology LLC A42275
Encompass_Preferred
Tipifarnib (also known as Zarnestra or R115777), an orally bioavailable quinolone analog of imidazole heterocyclics, is a potent and specific nonpeptidomimetic competitive inhibitor of farnesyltransferase (FTase). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4907