Learn More
Aviva Systems Biology Corp TIPIN Antibody - middle region (ARP50621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIPIN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TIPIN. Alternate names: FLJ20516ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60390