Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.8% | C9H12Cl2N4O2 | 25 MG
SDP
Catalog No. 5000358975
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research. It has antiangiogenic effects and inhibits the proliferation of endothelial cells. Tipiracil also acts as a potent inhibitor of thymidine phosphorylase, an enzyme that degrades FTD.
  • Thymidine phosphorylase inhibitor (TPI)
  • Used for cancer research
  • Antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Potent inhibitor of thymidine phosphorylase
  • Enzyme degrades FTD
Catalog No. 50-003-58975 Supplier Medchemexpress LLC Supplier No. 25MGHYA0063
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research. It has antiangiogenic effects and inhibits the proliferation of endothelial cells. Tipiracil also acts as a potent inhibitor of thymidine phosphorylase, an enzyme that degrades FTD.
  • Thymidine phosphorylase inhibitor (TPI)
  • Used for cancer research
  • Antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Potent inhibitor of thymidine phosphorylase
  • Enzyme degrades FTD