Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.9% | C9H12Cl2N4O2 | 10 MM 1 ML
SDP
Catalog No. 5000432074
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research.
  • Thymidine phosphorylase inhibitor
  • Suitable for cancer research
  • White to off-white solid appearance
  • Molecular weight: 279.12
  • Requires sealed storage at 4°C, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-32074 Supplier Medchemexpress LLC Supplier No. HYA006310MM/1ML
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research.
  • Thymidine phosphorylase inhibitor
  • Suitable for cancer research
  • White to off-white solid appearance
  • Molecular weight: 279.12
  • Requires sealed storage at 4°C, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month