Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.9% | C9H12Cl2N4O2 | 50 MG
SDP
Catalog No. 5000432125
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research. It exhibits antiangiogenic effects and inhibits the proliferation of endothelial cells. This compound is also a potent inhibitor of thymidine phosphorylase, the enzyme responsible for degrading FTD.
  • Thymidine phosphorylase inhibitor
  • Used in cancer research
  • Exhibits antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Potent inhibitor of thymidine phosphorylase
Catalog No. 50-004-32125 Supplier Medchemexpress LLC Supplier No. HYA006350MG
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research. It exhibits antiangiogenic effects and inhibits the proliferation of endothelial cells. This compound is also a potent inhibitor of thymidine phosphorylase, the enzyme responsible for degrading FTD.
  • Thymidine phosphorylase inhibitor
  • Used in cancer research
  • Exhibits antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Potent inhibitor of thymidine phosphorylase