Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.9% | C9H12Cl2N4O2 | 500 MG
SDP
Catalog No. 5000432124
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) primarily used in cancer research. It exhibits antiangiogenic effects and inhibits the proliferation of endothelial cells.
  • Inhibits thymidine phosphorylase
  • Antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Suitable for cancer research
Catalog No. 50-004-32124 Supplier Medchemexpress LLC Supplier No. HYA0063500MG
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) primarily used in cancer research. It exhibits antiangiogenic effects and inhibits the proliferation of endothelial cells.
  • Inhibits thymidine phosphorylase
  • Antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Suitable for cancer research