Learn More
TARGETMOL CHEMICALS INC TIPIRACIL HYDROCHLORIDE 25MG
SDP
Catalog No. 502577142
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Tipiracil hydrochloride (MA-1 hydrochloride) is a thymidine phosphorylase inhibitor (TPI). Tipiracil hydrochloride is one of the active components in TAS-102 which is an anticancer drug candidate currently in clinical trials. purity: 99%
Catalog No. 50-257-7142 Supplier TARGETMOL CHEMICALS INC Supplier No. T236625MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Tipiracil hydrochloride (MA-1 hydrochloride) is a thymidine phosphorylase inhibitor (TPI). Tipiracil hydrochloride is one of the active components in TAS-102 which is an anticancer drug candidate currently in clinical trials. purity: 99%