Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.9% | C9H12Cl2N4O2 | 5 MG
SDP
Catalog No. 501874003
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) primarily utilized in cancer research. It demonstrates antiangiogenic effects and effectively inhibits the proliferation of endothelial cells.
  • Potent inhibitor of thymidine phosphorylase
  • Used for cancer research
  • Exhibits antiangiogenic effects
  • Inhibits proliferation of endothelial cells
Catalog No. 50-187-4003 Supplier Medchemexpress LLC Supplier No. HYA00635MG
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) primarily utilized in cancer research. It demonstrates antiangiogenic effects and effectively inhibits the proliferation of endothelial cells.
  • Potent inhibitor of thymidine phosphorylase
  • Used for cancer research
  • Exhibits antiangiogenic effects
  • Inhibits proliferation of endothelial cells