Learn More
Cayman Chemical TIplaxtInIn 10mg
SDP
Catalog No. 502893984
Encompass_Preferred
A PAI-1 inhibitor (IC50 = 2.7 µM for the human enzyme); inhibits differentiation of human adipocytes at 5 µM; reduces tumor growth and microvessel density in a T24 bladder cancer mouse xenograft model at 5 and 20 mg/kg; prevents carotid artery occlusion, increases the time to occlusive thrombosis, and decreases the thrombus size in a rat model of ferric chloride-induced arterial thrombosis at 1 mg/kg
Catalog No. 50-289-3984 Supplier Cayman Chemical Supplier No. 900045010 MG
A PAI-1 inhibitor (IC50 = 2.7 µM for the human enzyme); inhibits differentiation of human adipocytes at 5 µM; reduces tumor growth and microvessel density in a T24 bladder cancer mouse xenograft model at 5 and 20 mg/kg; prevents carotid artery occlusion, increases the time to occlusive thrombosis, and decreases the thrombus size in a rat model of ferric chloride-induced arterial thrombosis at 1 mg/kg