Learn More
TARGETMOL CHEMICALS INC TIPLAXTININ 25MG
SDP
Catalog No. 502581038
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tiplaxtinin (Tiplasinin)(PAI-039) is a selective and orally efficacious inhibitor of plasminogen activator inhibitor-1 (PAI-1) with IC50 of 2.7 uM. purity: 99%
Catalog No. 50-258-1038 Supplier TARGETMOL CHEMICALS INC Supplier No. T203025MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tiplaxtinin (Tiplasinin)(PAI-039) is a selective and orally efficacious inhibitor of plasminogen activator inhibitor-1 (PAI-1) with IC50 of 2.7 uM. purity: 99%