Learn More
Apexbio Technology LLC Tiplaxtinin(PAI-039)(Synonyms: Tiplaxtinin, PAI-039, Tiplaxtinin (PAI-039), Tiplaxitin, PAI1 inhibitor, PAI-1 antagonist, RWJ-676070), 10mM (in 1mL DMSO), CAS: 393105-53-8.
SDP
Catalog No. 5000060646
Encompass_Preferred
Tiplaxtinin (PAI-039 CAS 393105-53-8) is an orally bioavailable small molecule inhibitor targeting plasminogen activator inhibitor-1 (PAI-1) a physiological regulator of the plasminogen activation system Elevated PAI-1 levels stabilize thrombi by inhibiting tissue- and urokinase-type plasminogen activators Tiplaxtinin binds selectively to PAI-1 with a Kd of approximately 480 nM and inhibits its activity with an IC50 value around 2 7 M In animal models oral administration of Tiplaxtinin significantly delayed arterial thrombus-induced occlusion supporting its utility as a research tool for studying fibrinolytic pathways and thrombotic disorders
Catalog No. 50-000-60646 Supplier Apexbio Technology LLC Supplier No. B59065.1
Tiplaxtinin (PAI-039 CAS 393105-53-8) is an orally bioavailable small molecule inhibitor targeting plasminogen activator inhibitor-1 (PAI-1) a physiological regulator of the plasminogen activation system Elevated PAI-1 levels stabilize thrombi by inhibiting tissue- and urokinase-type plasminogen activators Tiplaxtinin binds selectively to PAI-1 with a Kd of approximately 480 nM and inhibits its activity with an IC50 value around 2 7 M In animal models oral administration of Tiplaxtinin significantly delayed arterial thrombus-induced occlusion supporting its utility as a research tool for studying fibrinolytic pathways and thrombotic disorders