Learn More
Andwin Scientific TIPONE 0.1-10 5L NATURAL ELO
SDP
Supplier:  Andwin Scientific 11103700
Encompass_Preferred
TipOne 0.1-10/20 ul XL natural ultra micro pipet tips have a hydrophobic filter that are compsed of ultra high-density polyethylene. The filter traps aerosols and airborne contaminants in order to greatly reduce the risk of sample carry-over and cross contamination. This product contains improved 0.1-10/20 l XL ultra micro pipet tips fit most 0.1-2.5 l, 0.5-10 l, and 0.1-10 l pipettes. This product features improved graduation intervals: 1, 5, 10, and 20 l, holds up to 20 l, and has longer length to keep the tip collar outside of 1.5/2.0 ml tubes and most deep well plates, minimizing contamination. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-512