Learn More
Andwin Scientific TIPONE 0.1-10 5L NATURAL PIPE
SDP
Supplier:  Andwin Scientific 11113200
Encompass
TipOne 0.1-10 ul natural ultra micro pipet tips are available in a stacked format which saves space on the bench and on the storage shelf. The pipet tip's footprint is wider than most standard stacked tips to prevent overturning during use. Each tip layer locks into the one below it, preventing accidental separation. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 5 layers of two 96 tip wafers (960 total tips).
Catalog No. 50-819-554