Learn More
Andwin Scientific TIPONE 0.1-10 5L NATURAL PIPE
SDP
Supplier:  Andwin Scientific 11113000
Encompass_Preferred
TipOne 0.1-10 ul natural ultra micro pipet tips come in resealable bags containing 1000 tips. This product comes in 2.5 and 10 l graduations. This product comes in sealed bags with tamper evident strips and a resealable zipper closure. This product is certified free of RNase, DNase, DNA and pyrogens.
Catalog No. 50-819-553