Learn More
Andwin Scientific TIPONE .1-10 5L PIPET 960PK
SDP
Supplier:  Andwin Scientific 11213810
Encompass_Preferred
TipOne 0.1-10 l filter tips have a hydrophobic filter that are compsed of ultra high-density polyethylene. The filter traps aerosols and airborne contaminants in order to greatly reduce the risk of sample carry-over and cross contamination. This product is graduated at 2.5 and 10 l. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-579