Learn More
Andwin Scientific TIPONE 1-200 5L GRADUAT 4800PK
SDP
Supplier:  Andwin Scientific 11208860
Encompass_Preferred
TipOne 1-200 ul filter tips have a hydrophobic filter that are compsed of ultra high-density polyethylene. The filter traps aerosols and airborne contaminants in order to greatly reduce the risk of sample carry-over and cross contamination. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 50 racks of 96 tips (4800 total tips) with yellow color-coded wafers.
Catalog No. 50-819-578