Learn More
Andwin Scientific TIPONE 1-200 5L NATURAL 960PK
SDP
Supplier:  Andwin Scientific 11111200
Encompass_Preferred
TipOne 1-200 ul natural, beveled pipet tips are available in stacks which saves space on the bench and on the storage shelf. The pipet tip's footprint is wider than most standard stacked tips to prevent overturning during use. Each tip layer locks into the one below it, preventing accidental separation. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 5 layers of two 96 tip wafers (960 total tips).
Catalog No. 50-819-532