Learn More
Andwin Scientific TIPONE 1-200 5L NATURAL BEVE
SDP
Supplier:  Andwin Scientific 11101700
Encompass_Preferred
TipOne 1-200 ul Profile natural, graduated pipet tip refill wafers uses snap-in wafers that drop into an empty tip rack. The advanced tip design enhances fit and perfomance. Refills save more space and plastic. This product comes in 10, 50, and 100 l graduations.This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 wafers of 96 tips (960 total tips).
Catalog No. 50-819-505