Learn More
Andwin Scientific TIPONE 1-200 5L NATURAL BEVE
SDP
Supplier:  Andwin Scientific 11111810
Encompass_Preferred
TipOne 1-200 ul natural, beveled pipet tips come in sterilized racks. TipOne racks have a one-piece base that resists tipping during use. The hinged lid remains open at 120 for easy pipettor access. This product is compatible with multi-channel pipettes This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-541