Learn More
Andwin Scientific TIPONE 1-200 5L NATURAL PIPET
SDP
Supplier:  Andwin Scientific 11110800
Encompass_Preferred
TipOne 101-1000 ul natural, graduated pipet tips come in racks that have a contoured lid and base for easier handling. TipOne racks have a one-piece base that resists tipping during use. The hinged lid remains open at 120 for easy pipettor access. This product comes with raised rims on lid for easy stacking and storage. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-526