Learn More
Andwin Scientific TIPONE 101-1000 5L NATURAL G
SDP
Supplier:  Andwin Scientific 11112020
Encompass_Preferred
TipOne 101-1000 ul natural, graduated pipet tips come in resealable bags. The resealable bags come with tamper evident strips to ensure cleanliness. The included Profile Collar improves fit and the included Profile Wall is thinner and clearer than before. This product is graduated at 250, 500, and 1000 l. This product is certified free of RNase, DNase, DNA and pyrogens. This product contains 1000 tips.
Catalog No. 50-819-543