Learn More
Andwin Scientific TIPONE 101-1000 5L NATURAL G
SDP
Supplier:  Andwin Scientific 11112720
Encompass_Preferred
TipOne 101-1000 ul natural, graduated pipet tip refill wafers are graduated at 250, 500, and 1000 l. Refills save more space and plastic. The included Profile Collar improves fit and the included Profile Wall is thinner and clearer than before. This product uses snap-in wafers that drop into an empty tip rack. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-547