Learn More
CPC Scientific H-Tyr-Tic-Phe-Phe-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OPIO013A
Encompass
This peptide is a potent and highly selective antagonist at the delta-opioid receptor (IC50 = 6 nM) and therefore an ideal candidate for the study of opioid pharmacology (delta-opioid receptors).SEQUENCE: H-Tyr-Tic-Phe-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Y-Tic-FFMOLECULAR FORMULA: C37H38N4O6MOLECULAR WEIGHT: 634.73STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [146369-65-5]SYNONYMS: RESEARCH AREA: PainREFERENCES:P.W.Schiller et al., Proc. Natl. Acad. Sci. USA, 89, 11871 (1992)V.Kumar et al., J. Med. Chem., 43, 5050 (2000)D.Y.Maeda et al., J. Med. Chem., 43, 5044 (2000)SKU(s): OPIO-013A, OPIO-014
Catalog No. 50-210-9988