Learn More
TARGETMOL CHEMICALS INC TIPRANAVIR 5MG
SDP
Catalog No. 502579866
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tipranavir is a protease inhibitor that inhibits HIV-1 resistant to more than 1 protease inhibitor .Tipranavir effectively inhibits HIV-1 protease enzyme activity and dimerisation and has potent activity against multiple protease inhibitor (PI) HIV-1 isolates (IC50 of 66-410 nM). Tipranavir inhibits SARS-CoV-2 3CLpro activity. purity: 99%
Catalog No. 50-257-9866 Supplier TARGETMOL CHEMICALS INC Supplier No. T45785MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tipranavir is a protease inhibitor that inhibits HIV-1 resistant to more than 1 protease inhibitor .Tipranavir effectively inhibits HIV-1 protease enzyme activity and dimerisation and has potent activity against multiple protease inhibitor (PI) HIV-1 isolates (IC50 of 66-410 nM). Tipranavir inhibits SARS-CoV-2 3CLpro activity. purity: 99%