Learn More
Aviva Systems Biology Corp TIPRL Antibody - N-terminal region (ARP62048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIPRL. It was validated on Western Blot. . Alternate names: MGC3794, TIP, TIP41, dJ69E11.3, RP1-69E11.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-551