Learn More
Cayman Chemical Tipranavir-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1217819-15-2, 1 mg
SDP
Catalog No. 502940615
Encompass_Preferred
An internal standard for the quantification of tipranavir by GC- or LC-MS
Catalog No. 50-294-0615 Supplier Cayman Chemical Supplier No. 289071 MG
An internal standard for the quantification of tipranavir by GC- or LC-MS