Learn More
Thomas Scientific OneTouch e-dek Tips, 1000uL, Barrier, Paperboard Rack, Sterile
SDP
Catalog No. 501460659
Encompass
Tips are manufactured from the finest raw materials using precise production techniques that provide consistent quality performance. Universal fit on research grade pipettors and are great for multichannel pipettors. Graduated tips provide visible reference points with exceptional clarity. A wide selection of tips are available in multiple packaging options. All pipette tips are certified RNase/DNase-Free, Human DNA/PCR Inhibitor-Free, and non-pyrogenic.
Catalog No. 50-146-0659 Supplier Thomas Scientific Supplier No. 1165R39
Tips are manufactured from the finest raw materials using precise production techniques that provide consistent quality performance. Universal fit on research grade pipettors and are great for multichannel pipettors. Graduated tips provide visible reference points with exceptional clarity. A wide selection of tips are available in multiple packaging options. All pipette tips are certified RNase/DNase-Free, Human DNA/PCR Inhibitor-Free, and non-pyrogenic.