Learn More
CELLTREAT Scientific Products 1200µL Pipette Tip Reload System, LfTS Fit, Non-sterile
SDP
Catalog No. NC3377675
Encompass_Preferred
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tipsLfTS Reload Systems offer a convenient and economical way to refill empty tip racks, saving both time and moneyReload Systems dramatically reduce plastic waste by reusing empty racks10μL extended length tips have a maximum volume of 20μL and are compatible with 2, 10, and 20μL pipettesFine tip for improved accuracy and precision
Catalog No. NC3377675 Supplier CELLTREAT Scientific Products Supplier No. 229089
Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tipsLfTS Reload Systems offer a convenient and economical way to refill empty tip racks, saving both time and moneyReload Systems dramatically reduce plastic waste by reusing empty racks10μL extended length tips have a maximum volume of 20μL and are compatible with 2, 10, and 20μL pipettesFine tip for improved accuracy and precision