Learn More
CELLTREAT Scientific Products 300µL Pipette Tips, LfTS Fit, Racked, Sterile
SDP
Catalog No. NC2545512
Encompass_Preferred
Tips are designed to provide a more comfortable pipetting experiencePipette tips are packaged in racks with hinged lids designed for easy one-handed opening10μl tips feature an extended length for easier access to the bottom of vesselsFilter tips are fitted with a polymer aerosol barrier to reduce the risk of contaminationLids feature stacking supports, and are removable to save bench space
Catalog No. NC2545512 Supplier CELLTREAT Scientific Products Supplier No. 229083
Tips are designed to provide a more comfortable pipetting experiencePipette tips are packaged in racks with hinged lids designed for easy one-handed opening10μl tips feature an extended length for easier access to the bottom of vesselsFilter tips are fitted with a polymer aerosol barrier to reduce the risk of contaminationLids feature stacking supports, and are removable to save bench space