Learn More
MTC Bio 300µL pipette tips, sterile, for 200µL and 300µL pipettes, non- filtered, 10 racks of 96 tips
Catalog No. 501871082
Encompass_Preferred
These tips are designed to fit MTC’s exclusive 300µL single channel ProPette™ LE as well as most popular multi-channel pipettes.  Many researchers have recognized the need for an accurate pipette for volumes above 200µL, which is a range that is not well served by traditional 1mL pipettes. The unique 300µL ProPette™ LE and pipette tips close the gap and permit accurate and convenient pipetting, especially in the 200-300µL range. These tips also fit most popular 300µL multi-channel pipettes and are supplied pre-sterilized in 96 position racks, filtered or non-filtered.
Catalog No. 50-187-1082 Supplier MTC Bio Supplier No. P4300RK
These tips are designed to fit MTC’s exclusive 300µL single channel ProPette™ LE as well as most popular multi-channel pipettes.  Many researchers have recognized the need for an accurate pipette for volumes above 200µL, which is a range that is not well served by traditional 1mL pipettes. The unique 300µL ProPette™ LE and pipette tips close the gap and permit accurate and convenient pipetting, especially in the 200-300µL range. These tips also fit most popular 300µL multi-channel pipettes and are supplied pre-sterilized in 96 position racks, filtered or non-filtered.